![Anti-Gliadin (Wheat) antibody produced in rabbit fractionated antiserum, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/376/994/e2eb77fa-3715-4762-9965-24f5dd53607b/640/e2eb77fa-3715-4762-9965-24f5dd53607b.jpg)
Anti-Gliadin (Wheat) antibody produced in rabbit
![Anti-Gliadin (Wheat) antibody produced in rabbit fractionated antiserum, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/376/994/e2eb77fa-3715-4762-9965-24f5dd53607b/640/e2eb77fa-3715-4762-9965-24f5dd53607b.jpg)
Synonym(s):
Anti-Gliadin (Wheat)
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing