
4-Nitrobenzoic acid

Synonym(s):
4-Nitrobenzoic acid
Linear Formula:
O2NC6H4CO2H
CAS Number:
Molecular Weight:
167.12
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing