
4-Epianhydrotetracycline hydrochloride

Synonym(s):
(4R,4aS,12aS)-4-(Dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride, 4-Epianhydrotetracycline hydrochloride
Empirical Formula (Hill Notation):
C22H22N2O7 · HCl
CAS Number:
Molecular Weight:
462.88
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing