
2,3-Butanedithiol

Synonym(s):
2,3-Dimercaptobutane
Linear Formula:
CH3CH(SH)CH(SH)CH3
CAS Number:
Molecular Weight:
122.25
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing