Inorganic Elemental Impurity Mix Standards
![Pills Detection of elemental impurities in drug products as per ICH Q3D guidelines](/deepweb/assets/sigmaaldrich/marketing/global/images/categories/reference-materials/pills/pills.jpg)
Drug products may contain metal elemental impurities that present a risk to patient health. Elemental impurity levels must be monitored and should be below the daily intake levels set in the ICH Q3D guidelines.
In 2009, the International Conference on Harmonization (ICH) outlined a global policy for limiting metal impurities in drug products and ingredients. This approach required clear regulatory guidance on specification limits for elemental impurities and for national regulatory bodies to provide transparent and comparable results. The ICH classified elemental impurities in four different categories to facilitate decision making during risk assessment processes.
Beginning on January 1, 2018, the USP and the EP adopted ICH Q3D guidelines for elemental impurity limits. This resulted in an analytical method change for the determination of metal sulfide precipitation from colorimetric determination to analysis by ICP-OES or ICP-MS methods. We offer a wide selection of certified reference materials of different elemental impurity mixes and single elements with concentration limits in accordance ICHQ3D guidelines.
Section Overview
Elemental impurity mixes
Our products include standard mixes of TraceCERT® certified reference materials for different elements. They consist of element ratios corresponding to the oral, parenteral, and inhalation elemental concentration limits defined in ICHQ3D guidelines. They can be used for tests according to USP<232>, Ph. Eur. Gen. Chapter 5.20, as well as a mix for USP<2232> for dietary supplements.
- Products are made and analyzed under ISO/IEC 17025 and ISO 17034
- Product values are traceable to at least two independent references (NIST®, BAM, or SI unit kg)
- Convenient 100 mL pack sizes
- Packed in opaque bottles (FEP) and gas-tight, aluminum foil bags for extended stability
- Certificates provided in accordance with ISO Guide 31
Single element certified reference material solutions
For each classified and non-classified element, we offer single-element certified reference material solutions, under the Certipur® trademark, in addition to TraceCERT® brand, and NIST® SRM® selection.
- Suitable for inductively coupled plasma-optical emission spectrometry (ICP-OES) and inductively coupled plasma-mass spectrometry (ICP-MS) applications
- Available in multiple concentrations of 10 g/L, 1 g/L, 10 mg/L, and 1 mg/L.
- Tested for approximately 70 impurities
- Supplied in 100 mL HDPE or borosilicate glass bottles
Additionally, we offer multielement standard solutions in 100 mL HDPE bottles. The bottles are sealed in aluminum bags along with a certificate providing detailed documentation of uncertainty values, expiry date, and storage information.
Related Product Resources
- Analytix Reporter: Special Edition Pharma Analysis & GC
This special edition of the Analytix Reporter delves into pharmaceutical analysis and quality control, showcasing cutting-edge solutions, certified reference materials, and advanced methodologies for efficient impurity testing and comprehensive characterization of biologics and pharmaceuticals.
- Flyer: ICHQ3D
This informative flyer outlines the critical ICH Q3D guidelines for managing elemental impurities in pharmaceuticals, highlighting the importance of certified reference materials to ensure product safety and compliance in the drug development process.