Skip to Content
Merck
All Photos(1)

Key Documents

G005698

HPLC Analysis of Fat Soluble Vitamins on Ascentis® RP-Amide

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase: acetonitrile
flow rate: 2.0 mL/min
column temp.: 35 °C
detector: UV, 215 nm
injection: 20 μL

suitability

application for HPLC

application(s)

vitamins, nutraceuticals, and natural products

Analysis Note

The Ascentis RP-Amide is shown to be suitable for the analysis of fat soluble vitamins, alpha tocopherol (vitamin E) and cholecalciferol (vitamin D3 ).

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
    (±)-α-Tocopherol, analytical standard

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service