Skip to Content
Merck
All Photos(3)

Key Documents

03-249

Sigma-Aldrich

RIPAb+ Ago1 Antibody

clone 4G7-E12, from mouse

Synonym(s):

hAgo1, Argonaute1, eIF-2C 1, eIF2C 1, Q99, GERP95, DKFZp686M13167

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

biological source

mouse

Quality Level

clone

4G7-E12, monoclonal

species reactivity

human

manufacturer/tradename

RIPAb+
Upstate®

technique(s)

RIP: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

RIPAb+ antibodies are evaluated using the RNA Binding Protein Immunoprecipitation (RIP) assay. Each RIPAb+ antibody set includes a negative control antibody to ensure specificity of the RIP reaction and is verified for the co-immunoprecipitation of RNA associated specifically with the immunoprecipitated RNA-binding protein of interest. Where appropriate, the RIPAb+ set also includes quantitative RT-PCR control primers (RIP Primers) to biologically validate your IP results by successfully co-precipitating the specific RNA targets, such as messenger RNAs. The qPCR protocol and primer sequences are provided, allowing researchers to validate RIP protocols when using the antibody in their experimental context. If a target specific assay is not provided, the RIPAb+ kit is validated using an automated microfluidics-based assay by enrichment of detectable RNA over control immunoprecipitation.
Ago1 (Eukaryotic translation initiation factor 2C 1) is a member of the argonaute family. It contains a PAZ and a PIWI domain, and is required for G1 arrest and mating in response to nitrogen starvation. It may also contribute to cell cycle control by regulating cytokinesis. Ago1 is also needed for the production of mature miRNA that affects miRNA-directed RNA cleavage and is therefore important for heterchromatin formation and accurate chromosome segregation.

Specificity

This antibody has been shown by an independent laboratory to not cross react with Ago2, Ago3, or Ago4 (Azuma-Mukai A et al. (2008) Proc. Natl. Acad. Sci. USA 105: 7964-7969).

Immunogen

GST-tagged recombinant protein corresponding to human Ago1

Application

RNA Binding Protein Immunoprecipitation (microRNA):
Representative lot data.
RIP Lysate prepared from 293 cells (2 X 107 cell equivalents per IP) were subjected to immunoprecipitation using 5 µg of either a normal mouse IgG (Cat. # CS200621), or Anti-Ago1 antibody (Cat. # CS207365) and the Magna RIP RNA-Binding Protein Immunoprecipitation Kit (Cat. # 17-700).
Successful immunoprecipitation of Ago 1-associated microRNA was verified by TaqMan microRNA assay, hsa-miR-15a. (Figure2). Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.
Western Blot Analysis:
Representative lot data.
HeLa cell lysate was probed with Anti-Ago 1, clone 4G7-E12 (0.1 µg/mL). Proteins were visualized using a Goat Anti-Mouse IgG secondary antibody conjugated to HRP and a chemiluminescence detection system.
Arrow indicates Ago 1 (~97 kDa).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Binding Protein (RBP)
This RIPAb+ Ago1 -RIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

10 assays per set. Recommended use: ~5 µg of antibody per RIP (dependent upon biological context).

Quality

RNA Binding Protein Immunoprecipitation:
Representative lot data
RIP Lysate prepared from 293 cells (2 X 10E7 cell equivalents per IP) were subjected to immunoprecipitation using 5 µg of either a normal mouse IgG, or 5 µg Anti-Ago1 antibody and the Magna RIP® RNA-Binding Protein Immunoprecipitation Kit (Cat. # 17-700).
Successful immunoprecipitation of Argonaute 1-associated RNA was verified by qPCR using RIP Primers IGF2, (Figure 1).
Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.

Target description

~97 kDa

Physical form

Anti-Ago1 (Mouse Monoclonal). One vial containing 50 µg protein G purified monoclonal in buffer containing 0.1 M Tris-glycine (pH 7.4), 150mM NaCl, 0.05% sodium azide before the addition of glycerol to 30%. Store at -20° C.
Concentration: 0.7 mg/mL
Normal mouse IgG. One vial containing 125 µg purified mouse IgG in 125 µL storage buffer containing 0.1% sodium azide. Store at -20°C.
RIP Primer IGF2. One vial containing 75 μL of 5 μM of each primer specific for human IGF2 mRNA. Store at -20°C.
FOR: GCG GCT TCT ACT TCA GCA G
REV: CAG GTG TCA TAT TGG AAG AAC
Format: Purified
Protein G Purified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Includes normal mouse IgG and primers specific for human IGF2 mRNA.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MAGNA RIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingwei Cao et al.
Cancer cell international, 20, 327-327 (2020-07-24)
Long non-coding RNAs (lncRNAs) has been extensively reported play important roles in regulating the development and progression of cancers, including Glioblastoma (GBM). LINC01426 is a novel lncRNA that has been identified as an oncogenic gene in GBM. Herein, we attempted
Teresa Bellissimo et al.
Cell death & disease, 10(1), 17-17 (2019-01-10)
To perform their regulatory functions, microRNAs (miRNAs) must assemble with any of the four mammalian Argonaute (Ago) family of proteins, Ago1-4, into an effector complex known as the RNA-induced silencing complex (RISC). While the mature miRNA guides the RISC complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service