Key Documents
TEM-LC300AUUL
Sigma-Aldrich
Ultrathin Lacey Carbon Supported Gold Grid
size 300 mesh, box of 25
Synonym(s):
LACEY CARBON FILM - AU 300 MESH ULTRA THIN 25/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
size
300 mesh
General description
Ultrathin Lacey Carbon (4-6 nm) Supported Grid: Gold Grid, 300 mesh. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.