HPLC Analysis of Ketoconazole Enantiomers on Astec® CYCLOBOND I 2000 HP-RSP
CONDITIONS
column
CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 μm particles (24024AST)
mobile phase
[A] 0.1% formic acid; [B] acetonitrile; (70:30, A:B)
flow rate
1 mL/min
column temp.
25 °C
injection
1 μL
detector
UV, 230 nm
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null