GC Analysis of Lemon Essential Oil on SLB®-IL59
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL59, 30 m x 0.25 mm I.D., 0.20 μm (28891-U)
oven
50 °C, 5 °C/min to 300 °C
inj. temp.
250 °C
detector
FID, 305 °C
carrier gas
helium, 40 cm/sec
injection
1 μL 100:1 split
sample
lemon essential oil diluted 1:10 in hexane
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany