GC Analysis of Ionic Liquid Test Mix on SLB®-IL59 (30 m x 0.25 mm I.D., 0.20 μm)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL59, 30 m x 0.25 mm I.D., 0.20 μm (28891-U)
oven
110 °C
inj. temp.
110 °C
detector
FID, 250 °C
carrier gas
helium, 26 cm/sec
injection
1.0 μL 100:1 split
sample
Ionic Liquid Test Mix #1, each analyte at various concentrations in isooctane (N09000)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany