Merck
  • Formation of environmentally relevant polyhalogenated carbazoles from chloroperoxidase-catalyzed halogenation of carbazole.
Formation of environmentally relevant polyhalogenated carbazoles from chloroperoxidase-catalyzed halogenation of carbazole.
Environmental pollution (Barking, Essex : 1987) (2017-09-28)
Yanqiu Chen, Kunde Lin, Da Chen, Kun Wang, Wenxiu Zhou, Yan Wu, Xinwen Huang
ABSTRACT
Polyhalogenated carbazoles (PHCs) are a class of emerging organic contaminants that have received increasing concern due to their widespread distribution and dioxin-like toxicity. Although previous studies have suggested possible natural sources of PHCs in the environment, the formation pathways are poorly understood. Here we explored the production of PHCs from halogenation of carbazole in the presence of Br
Sigma-Aldrich
Carbazole, ≥95% (GC)
Sigma-Aldrich
3,6-Dibromocarbazole, 97%