Merck
Nitric oxide donor [Ru(terpy)(bdq)NO]
Free radical biology & medicine (2017-09-14)
Simone R Potje, Zhenlong Chen, Suellen D'Arc S Oliveira, Lusiane M Bendhack, Roberto S da Silva, Marcelo G Bonini, Cristina Antoniali, Richard D Minshall
ABSTRACT
[Ru(terpy)(bdq)NO]
Sigma-Aldrich
Superoxide dismutase–polyethylene glycol from bovine erythrocytes, lyophilized powder, ≥1,350 units/mg protein