Merck
  • The pivotal role of astrocytes in an in vitro stroke model of the blood-brain barrier.
The pivotal role of astrocytes in an in vitro stroke model of the blood-brain barrier.
Frontiers in cellular neuroscience (2014-11-13)
Winfried Neuhaus, Fabian Gaiser, Anne Mahringer, Jonas Franz, Christoph Riethmüller, Carola Förster
ABSTRACT
Stabilization of the blood-brain barrier during and after stroke can lead to less adverse outcome. For elucidation of underlying mechanisms and development of novel therapeutic strategies validated in vitro disease models of the blood-brain barrier could be very helpful. To mimic in vitro stroke conditions we have established a blood-brain barrier in vitro model based on mouse cell line cerebEND and applied oxygen/glucose deprivation (OGD). The role of astrocytes in this disease model was investigated by using cell line C6. Transwell studies pointed out that addition of astrocytes during OGD increased the barrier damage significantly in comparison to the endothelial monoculture shown by changes of transendothelial electrical resistance as well as fluorescein permeability data. Analysis on mRNA and protein levels by qPCR, western blotting and immunofluorescence microscopy of tight junction molecules claudin-3,-5,-12, occludin and ZO-1 revealed that their regulation and localisation is associated with the functional barrier breakdown. Furthermore, soluble factors of astrocytes, OGD and their combination were able to induce changes of functionality and expression of ABC-transporters Abcb1a (P-gp), Abcg2 (bcrp), and Abcc4 (mrp4). Moreover, the expression of proteases (matrixmetalloproteinases MMP-2, MMP-3, MMP-9, and t-PA) as well as of their endogenous inhibitors (TIMP-1, TIMP-3, PAI-1) was altered by astrocyte factors and OGD which resulted in significant changes of total MMP and t-PA activity. Morphological rearrangements induced by OGD and treatment with astrocyte factors were confirmed at a nanometer scale using atomic force microscopy. In conclusion, astrocytes play a major role in blood-brain barrier breakdown during OGD in vitro.
Sigma-Aldrich
Ko143 hydrate, ≥98% (HPLC)
Sigma-Aldrich
MK-571 sodium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Roche
cOmplete ULTRA Tablets, Mini, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.