Merck
  • Simultaneous determination of size and quantification of silica nanoparticles by asymmetric flow field-flow fractionation coupled to ICPMS using silica nanoparticles standards.
Simultaneous determination of size and quantification of silica nanoparticles by asymmetric flow field-flow fractionation coupled to ICPMS using silica nanoparticles standards.
Analytical chemistry (2015-01-30)
Francisco Barahona, Otmar Geiss, Patricia Urbán, Isaac Ojea-Jimenez, Douglas Gilliland, Josefa Barrero-Moreno
ABSTRACT
This work proposes the use of multimodal mixtures of monodispersed silica nanoparticles (SiO2-NPs) standards for the simultaneous determination of size and concentration of SiO2-NPs in aqueous suspensions by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass spectrometry (ICPMS). For such a purpose, suspensions of SiO2-NPs standards of 20, 40, 60, 80, 100, and 150 nm were characterized by transmission electronic microscopy (TEM), centrifugal liquid sedimentation (CLS), dynamic light scattering (DLS) and by measuring the Z-potential of the particles as well as the exact concentration of NPs by offline ICPMS. An online AF4-ICPMS method which allowed the separation of all the different sized SiO2-NPs contained in the mixture of standards was developed and the analytical figures of merit were systematically evaluated. The method showed excellent linearity in the studied concentration range (0.1-25 mg L(-1)), limits of detection between 0.16 and 0.3 mg L(-1) for smaller and greater particles, respectively, besides a satisfactory accuracy. AF4 calibration with particles with identical nature to those to be analyzed, also permitted accurate size determination in a pragmatic way. Similarly, by using prechannel calibration with NPs for mass determination it was possible to overcome common quantification problems associated with losses of material during the separation and size-dependent effects. The proposed methodology was successfully applied to the characterization in terms of size and concentration of aqueous test samples containing SiO2-NPs with monomodal size distributions.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium carbonate, for inorganic trace analysis, 99.999% trace metals basis