Merck
  • Determination of phosphate in serum and urine by a single step malachite-green method.
Determination of phosphate in serum and urine by a single step malachite-green method.
Clinica chimica acta; international journal of clinical chemistry (1975-02-22)
A Kallner
ABSTRACT
The highly sensitive malachite-green method for the determination of phosphate requires a protective colloid in order to avoid precipitation of the formed dye salt. A polyvinylalcohol proved to be suitable and permit the determination of phosphate in urine and in serum without prior precipitation of the proteins. The present method is suitable for a one-step procedure which allows the analysis of several hundred samples per man-hour and is applicable to microquantities of serum or urine.
Sigma-Aldrich
Malachite Green oxalate salt, crystalline, S. No.: 754