Merck
  • In position 7 L- and D-Tic-substituted oxytocin and deamino oxytocin: NMR study and conformational insights.
In position 7 L- and D-Tic-substituted oxytocin and deamino oxytocin: NMR study and conformational insights.
Amino acids (2010-01-29)
Zinovia Spyranti, Maria Fragiadaki, Vassiliki Magafa, Lenka Borovickova, Georgios A Spyroulias, Paul Cordopatis, Jirina Slaninova
ABSTRACT
Incorporation of L- or D-Tic into position 7 of oxytocin (OT) and its deamino analogue ([Mpa(1)]OT) resulted in four analogues, [L-Tic(7)]OT (1), [D-Tic(7)]OT (2), [Mpa(1),L-Tic(7)]OT (3) and [Mpa(1),D-Tic(7)]OT (4). Their biological properties were described by Fragiadaki et al. (Eur J Med Chem 42:799-806, 2007). Their NMR study (NOESY, TOCSY, (1)H-(13)C HSQC spectra) is presented here. Analogues 1, 3 and 4 showed partial agonistic activity, analogue 2 was pure antagonist, suggesting that a cis conformation between residues 6 and 7 of the molecule does not result in antagonistic activity. However, the reduction in agonistic activity of analogues 1, 3 and 4 in comparison to oxytocin is consistent with the reduction of the trans conformation form. Binding affinity for the human oxytocin receptor with IC(50) value of 130, 730, 103, and 380 nM for peptides 1, 2, 3, and 4, respectively, showed lower affinity in the case of D analogues. Deamination slightly increased the affinity. The existence of both cis and trans configurations of the Cys(6)-D-Tic(7) bond is supported by observation of two sets of cross-peaks for (1)H and (13)C nuclei for most of the residues of the peptide not only in NOESY and TOCSY but also in (1)H-(13)C HSQC spectra. The MS and HPLC indicate the presence of a single molecule/peptide, and NMR data thus suggest that this second set of peaks is due to the cis conformation.
Sigma-Aldrich
1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride, 96%
Sigma-Aldrich
(S)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid, 97%