Merck
  • Concomitant facilitation of GABAA receptors and KV7 channels by the non-opioid analgesic flupirtine.
Concomitant facilitation of GABAA receptors and KV7 channels by the non-opioid analgesic flupirtine.
British journal of pharmacology (2011-12-23)
Felicia Klinger, Petra Geier, Mario M Dorostkar, Giri K Chandaka, Arsalan Yousuf, Isabella Salzer, Helmut Kubista, Stefan Boehm
ABSTRACT
Flupirtine is a non-opioid analgesic that has been in clinical use for more than 20 years. It is characterized as a selective neuronal potassium channel opener (SNEPCO). Nevertheless, its mechanisms of action remain controversial and are the purpose of this study. Effects of flupirtine on native and recombinant voltage- and ligand-gated ion channels were explored in patch-clamp experiments using the following experimental systems: recombinant K(IR)3 and K(V)7 channels and α3β4 nicotinic acetylcholine receptors expressed in tsA 201 cells; native voltage-gated Na(+), Ca(2+), inward rectifier K(+), K(V)7 K(+), and TRPV1 channels, as well as GABA(A), glycine, and ionotropic glutamate receptors expressed in rat dorsal root ganglion, dorsal horn and hippocampal neurons. Therapeutic flupirtine concentrations (≤10 µM) did not affect voltage-gated Na(+) or Ca(2+) channels, inward rectifier K(+) channels, nicotinic acetylcholine receptors, glycine or ionotropic glutamate receptors. Flupirtine shifted the gating of K(V)7 K(+) channels to more negative potentials and the gating of GABA(A) receptors to lower GABA concentrations. These latter effects were more pronounced in dorsal root ganglion and dorsal horn neurons than in hippocampal neurons. In dorsal root ganglion and dorsal horn neurons, the facilitatory effect of therapeutic flupirtine concentrations on K(V)7 channels and GABA(A) receptors was comparable, whereas in hippocampal neurons the effects on K(V)7 channels were more pronounced. These results indicate that flupirtine exerts its analgesic action by acting on both GABA(A) receptors and K(V)7 channels.
Sigma-Aldrich
Flupirtine maleate salt, ≥98% (HPLC)