Merck
  • SnO2@graphene nanocomposites as anode materials for Na-ion batteries with superior electrochemical performance.
SnO2@graphene nanocomposites as anode materials for Na-ion batteries with superior electrochemical performance.
Chemical communications (Cambridge, England) (2013-03-13)
Dawei Su, Hyo-Jun Ahn, Guoxiu Wang
ABSTRACT
An in situ hydrothermal synthesis approach has been developed to prepare SnO2@graphene nanocomposites. The nanocomposites exhibited a high reversible sodium storage capacity of above 700 mA h g(-1) and excellent cyclability for Na-ion batteries. In particular, they also demonstrated a good high rate capability for reversible sodium storage.
Sigma-Aldrich
Tin(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, nanopowder, ≤100 nm avg. part. size