Merck
  • Nocardioides perillae sp. nov., isolated from surface-sterilized roots of Perilla frutescens.
Nocardioides perillae sp. nov., isolated from surface-sterilized roots of Perilla frutescens.
International journal of systematic and evolutionary microbiology (2012-06-19)
Hui-Jing Du, Yu-Zhen Wei, Jing Su, Hong-Yu Liu, Bai-Ping Ma, Bao-Lin Guo, Yu-Qin Zhang, Li-Yan Yu
ABSTRACT
A Gram-stain-positive, rod-shaped actinobacterium, designated strain I10A-01402(T), was isolated from surface-sterilized roots of a medicinal plant, Perilla frutescens, collected in a suburb of Beijing, China. Chemotaxonomically, the strain contained ll-diaminopimelic acid as the diagnostic diamino acid and MK-8(H4) as the predominant menaquinone. The phospholipids were diphosphatidylglycerol, phosphatidylglycerol and phosphatidylinositol. The major fatty acids were C17 : 1ω9c, C18 : 1ω9c, C17 : 0, C16 : 0 and iso-C16 : 0. The genomic DNA G+C content was 70.4 mol%. 16S rRNA gene sequence analysis indicated that strain I10A-01402(T) belonged to the genus Nocardioides. Phylogenetic analyses based on 16S rRNA gene sequences showed that the isolate formed a robust cluster with Nocardioides ginsengisegetis Gsoil 485(T), N. koreensis MSL-09(T) and N. alkalitolerans KSL-1(T). On the basis of the evidence from our polyphasic taxonomic study, a novel species, Nocardioides perillae sp. nov., is proposed. The type strain is I10A-01402(T) ( = CPCC 203382(T)  = DSM 24552(T)  = KCTC 29022(T)).
Sigma-Aldrich
2,6-Diaminopimelic acid, ≥98% (TLC)
Sigma-Aldrich
2,6-Diaminopimelic acid, ≥95.0% (NT)