Merck
  • Residues characterisation from the fluidised bed combustion of East London's solid recovered fuel.
Residues characterisation from the fluidised bed combustion of East London's solid recovered fuel.
Waste management (New York, N.Y.) (2010-03-17)
D E Balampanis, S J T Pollard, N Simms, P Longhurst, F Coulon, R Villa
ABSTRACT
Waste thermal treatment in Europe is moving towards the utilisation of the combustible output of mechanical, biological treatment (MBT) plants. The standardisation of solid recovered fuels (SRF) is expected to support this trend and increase the amount of the generated combustion residues. In this work, the residues and especially the fly ashes from the fluidised bed combustion (FBC) of East London's NCV 3, Cl 2, and Hg 1 class SRF, are characterised. The following toxicity indicators have been studied: leachable chlorine, organochlorides expressed as pentachlorobenzene and hexachlorobenzene, and the heavy metals Cu, Cr, Cd, Zn, Ni, and Pb. Furthermore the mineralogical pattern of the ashes has been studied by means of XRD and SEM-EDS. The results suggest that these SRF derived ashes have significantly lower quantities of Cu, Cd, Pb, Zn, leachable Cl, and organochlorides when compared to other literature values from traditional waste thermal treatment applications. This fact highlights the importance of modern separation technologies employed in MBT plants for the removal of components rich in metals and chlorine from the combustible output fraction of SRF resulting to less hazardous residues.
Product Description
Supelco
Pentachlorobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Pentachlorobenzene, 96%