Merck
  • Pharmacokinetic, metabolism and withdrawal time of orphenadrine in camels (Camelus dromedarius) after intravenous administration.
Pharmacokinetic, metabolism and withdrawal time of orphenadrine in camels (Camelus dromedarius) after intravenous administration.
Research in veterinary science (2008-03-07)
M Elghazali, I A Wasfi, A A Abdel Hadi, A M Latum
ABSTRACT
The pharmacokinetics of orphenadrine (ORPH) following a single intravenous (i.v.) dose was investigated in six camels (Camelus dormedarius). Orphenadrine was extracted from the plasma using a simple sensitive liquid-liquid extraction method and determined by gas chromatography/mass spectrometry (GC/MS). Following i.v. administration plasma concentrations of ORPH decline bi-exponentially with distribution half-life (t(1/2)(alpha)) of 0.50+/-0.07h, elimination half-life (t(1/2)(beta)) of 3.57+/-0.55h, area under the time concentration curve (AUC) of 1.03+/-0.10g/hl(-1). The volume of distribution at steady state (Vd(ss)) 1.92+/-0.22lkg(-1), volume of the central compartment of the two compartment pharmacokinetic model (V(c)) 0.87+/-0.09lkg(-1), and total body clearance (Cl(T)) of 0.60+/-0.09l/hkg(-1). Three orphenadrine metabolites were identified in urine samples of camels. The first metabolite N-desmethyl-orphenadrine resulted from N-dealkylation of ORPH with molecular ion m/z 255. The second N,N-didesmethyl-orphenadrine, resulted from N-didesmethylation with molecular ion m/z 241. The third metabolite, hydroxyl-orphenadrine, resulted from the hydroxylation of ORPH with molecular ion m/z 285. ORPH and its metabolites in camel were extensively eliminated in conjugated form. ORPH remains detectable in camel urine for three days after i.v. administration of a single dose of 350mg orphenadrine aspartate.
Product Description
Supelco
Orphenadrine citrate salt, analytical standard
Orphenadrine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Orphenadrine hydrochloride, ≥98.0% (AT)