Merck
  • G1/S arrest induced by histone deacetylase inhibitor sodium butyrate in E1A + Ras-transformed cells is mediated through down-regulation of E2F activity and stabilization of beta-catenin.
G1/S arrest induced by histone deacetylase inhibitor sodium butyrate in E1A + Ras-transformed cells is mediated through down-regulation of E2F activity and stabilization of beta-catenin.
The Journal of biological chemistry (2006-05-24)
Maria V Abramova, Tatiana V Pospelova, Fedor P Nikulenkov, Christine M Hollander, Albert J Fornace, Valery A Pospelov
ABSTRACT
Tumor cells are often characterized by a high and growth factor-independent proliferation rate. We have previously shown that REF cells transformed with oncogenes E1A and c-Ha-Ras do not undergo G(1)/S arrest of the cell cycle after treatment with genotoxic factors. In this work, we used sodium butyrate, a histone deacetylase inhibitor, to show that E1A + Ras transformants were able to stop proliferation and undergo G(1)/S arrest. Apart from inducing G(1)/S arrest, sodium butyrate was shown to change expression of a number of cell cycle regulatory genes. It down-regulated cyclins D1, E, and A as well as c-myc and cdc25A and up-regulated the cyclin-kinase inhibitor p21(waf1). Accordingly, activities of cyclin E-Cdk2 and cyclin A-Cdk2 complexes in sodium butyrate-treated cells were decreased substantially. Strikingly, E2F1 expression was also down-modulated at the levels of gene transcription, the protein content, and the E2F transactivating capability. To further study the role of p21(waf1) in the sodium butyrate-induced G(1)/S arrest and the E2F1 down-modulation, we established E1A + Ras transformants from mouse embryo fibroblast cells with deletion of the cdkn1a (p21(waf1)) gene. Despite the absence of p21(waf1), sodium butyrate-treated mERas transformants reveal a slightly delayed G(1)/S arrest as well as down-modulation of E2F1 activity, implying that the observed effects are mediated through an alternative p21(waf1)-independent signaling pathway. Subsequent analysis showed that sodium butyrate induced accumulation of beta-catenin, a downstream component of the Wnt signaling. The results obtained indicate that the antiproliferative effect of histone deacetylase inhibitors on E1A + Ras-transformed cells can be mediated, alongside other mechanisms, through down-regulation of E2F activity and stabilization of beta-catenin.
Sigma-Aldrich
Isobutyric acid, puriss. p.a., ≥99.5%
Supelco
Isobutyric acid, analytical standard
Sigma-Aldrich
Isobutyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Isobutyric acid, ≥99%, FCC, FG
Sigma-Aldrich
Isobutyric acid, 99%