Merck
  • 13C and 2H kinetic isotope effects and the mechanism of bromination of 1-pentene under synthetic conditions.
13C and 2H kinetic isotope effects and the mechanism of bromination of 1-pentene under synthetic conditions.
Organic letters (2000-05-24)
S R Merrigan, D A Singleton
ABSTRACT
[formula: see text] The 13C and 2H kinetic isotope effects for the bromination of 1-pentene with Br2 in CCl4 were determined and interpreted with the aid of calculationally predicted isotope effects. The isotope effects observed are consistent with rate-limiting bromonium ion formation and do not fit with either rate-limiting production of a pi complex or reaction of a reversibly formed bromonium ion. This rules out some of the mechanistic complexities suggested for other brominations, though the identity of the brominating reagent(s) under these synthetic conditions remains uncertain.
Sigma-Aldrich
1-Pentene, 98%
Sigma-Aldrich
1-Pentene, ≥98.5% (GC)
Supelco
1-Pentene, analytical standard