Merck
  • Insulin-like growth factor-binding protein-7 (IGFBP7) links senescence to heart failure.
Insulin-like growth factor-binding protein-7 (IGFBP7) links senescence to heart failure.
Nature cardiovascular research (2022-12-01)
Liyong Zhang, David Smyth, Mohammad Al-Khalaf, Alice Blet, Qiujiang Du, Jordan Bernick, Michael Gong, Xu Chi, Yena Oh, Malaika Roba-Oshin, Elizabeth Coletta, Michel Feletou, Anthony O Gramolini, Kyoung-Han Kim, Thais Coutinho, James L Januzzi, Benoit Tyl, Andre Ziegler, Peter P Liu
ABSTRACT
Heart failure (HF) is a rising global cardiovascular epidemic driven by aging and chronic inflammation. As elderly populations continue to increase, precision treatments for age-related cardiac decline are urgently needed. Here we report that cardiac and blood expression of IGFBP7 is robustly increased in patients with chronic HF and in an HF mouse model. In a pressure overload mouse HF model, Igfbp7 deficiency attenuated cardiac dysfunction by reducing cardiac inflammatory injury, tissue fibrosis and cellular senescence. IGFBP7 promoted cardiac senescence by stimulating IGF-1R/IRS/AKT-dependent suppression of FOXO3a, preventing DNA repair and reactive oxygen species (ROS) detoxification, thereby accelerating the progression of HF. In vivo, AAV9-shRNA-mediated cardiac myocyte Igfbp7 knockdown indicated that myocardial IGFBP7 directly regulates pathological cardiac remodeling. Moreover, antibody-mediated IGFBP7 neutralization in vivo reversed IGFBP7-induced suppression of FOXO3a, restored DNA repair and ROS detoxification signals and attenuated pressure-overload-induced HF in mice. Consequently, selectively targeting IGFBP7-regulated senescence pathways may have broad therapeutic potential for HF.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Sigma-Aldrich
Anti-STING Antibody, clone S17G2C4B7, clone S17G2C4B7, from mouse
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Anti-IRS1 Antibody, Upstate®, from rabbit