Merck
  • Synthetic analogues of the montanine-type alkaloids with activity against apoptosis-resistant cancer cells.
Synthetic analogues of the montanine-type alkaloids with activity against apoptosis-resistant cancer cells.
Bioorganic & medicinal chemistry letters (2018-02-08)
Karthik Govindaraju, Aude Ingels, Md Nabiul Hasan, Dandan Sun, Veronique Mathieu, Marco Masi, Antonio Evidente, Alexander Kornienko
ABSTRACT
In a search of small molecules active against apoptosis-resistant cancer cells, a skeletal rearrangement of alkaloid haemanthamine was utilized to generate a series of compounds possessing the alkaloid montanine ring system. The synthesized compounds were found to inhibit proliferation of cancer cells resistant to apoptosis at micromolar concentrations. Selected compounds were also active against patient-derived glioblastoma cells expressing stem-cell markers. This is the first report describing the preparation of synthetic analogues of the montanine-type alkaloids with antiproliferative activity. The compounds prepared in the current investigation appear to be a useful starting point for the development of agents to fight cancers with apoptosis resistance, and thus, associated with poor prognoses.
Product Description
U-373 MG (Uppsala) cell line, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.