Merck
  • Retinoic Acid Receptor-β Is Downregulated in Hepatocellular Carcinoma and Cirrhosis and Its Expression Inhibits Myosin-Driven Activation and Durotaxis in Hepatic Stellate Cells.
Retinoic Acid Receptor-β Is Downregulated in Hepatocellular Carcinoma and Cirrhosis and Its Expression Inhibits Myosin-Driven Activation and Durotaxis in Hepatic Stellate Cells.
Hepatology (Baltimore, Md.) (2018-07-29)
Ernesto Cortes, Dariusz Lachowski, Alistair Rice, Antonios Chronopoulos, Benjamin Robinson, Stephen Thorpe, David A Lee, Lucia A Possamai, Haiyun Wang, David J Pinato, Armando E Del Río Hernández
ABSTRACT
Hepatic stellate cells (HSCs) are essential perisinusoidal cells in both healthy and diseased liver. HSCs modulate extracellular matrix (ECM) homeostasis when quiescent, but in liver fibrosis, HSCs become activated and promote excess deposition of ECM molecules and tissue stiffening via force generation and mechanosensing. In hepatocellular carcinoma (HCC), activated HSCs infiltrate the stroma and migrate to the tumor core to facilitate paracrine signaling with cancer cells. Because the function of HSCs is known to be modulated by retinoids, we investigated the expression profile of retinoic acid receptor beta (RAR-β) in patients with cirrhosis and HCC, as well as the effects of RAR-β activation in HSCs. We found that RAR-β expression is significantly reduced in cirrhotic and HCC tissues. Using a comprehensive set of biophysical methods combined with cellular and molecular biology, we have elucidated the biomechanical mechanism by which all trans-retinoic acid promotes HSC deactivation via RAR-β-dependent transcriptional downregulation of myosin light chain 2 expression. Furthermore, this also abrogated mechanically driven migration toward stiffer substrates. Conclusion: Targeting mechanotransduction in HSCs at the transcriptional level may offer therapeutic options for a range of liver diseases.
Sigma-Aldrich
Anti-MLC-2 Antibody, clone 19D3.1, clone 19D3.1, from mouse
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%