Merck
  • Baseline iron status and presence of anaemia determine the course of systemic Salmonella infection following oral iron supplementation in mice.
Baseline iron status and presence of anaemia determine the course of systemic Salmonella infection following oral iron supplementation in mice.
EBioMedicine (2021-09-07)
Alexander Hoffmann, David Haschka, Lara Valente de Souza, Piotr Tymoszuk, Markus Seifert, Laura von Raffay, Richard Hilbe, Verena Petzer, Patrizia L Moser, Manfred Nairz, Günter Weiss
ABSTRACT
Iron deficiency anaemia (IDA) is a major health concern. However, preventive iron supplementation in regions with high burden of infectious diseases resulted in an increase of infection related morbidity and mortality. We fed male C57BL/6N mice with either an iron deficient or an iron adequate diet. Next, they received oral iron supplementation or placebo followed by intraperitoneal infection with Salmonella Typhimurium (S.Tm). We found that mice with IDA had a poorer clinical outcome than mice on an iron adequate diet. Interestingly, iron supplementation of IDA mice resulted in higher bacterial burden in organs and shortened survival. Increased transferrin saturation and non-transferrin bound iron in the circulation together with low expression of ferroportin facilitated the access of the pathogen to iron and promoted bacterial growth. Anaemia, independent of iron supplementation, was correlated with reduced neutrophil counts and cytotoxic T cells. With iron supplementation, anaemia additionally correlated with increased splenic levels of the cytokine IL-10, which is suggestive for a weakened immune control to S.Tm infection. Supplementing iron to anaemic mice worsens the clinical course of bacterial infection. This can be traced back to increased iron delivery to bacteria along with an impaired anti-microbial immune response. Our findings may have important implications for iron supplementation strategies in areas with high endemic burden of infections, putting those individuals, who potentially profit most from iron supplementation for anaemia, at the highest risk for infections. Financial support by the Christian Doppler Laboratory for Iron Metabolism and Anemia Research.
Sigma-Aldrich
Anti-MAP3K5 (Ab-83) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution