Merck
  • Sildenafil (Viagra) Aggravates the Development of Experimental Abdominal Aortic Aneurysm.
Sildenafil (Viagra) Aggravates the Development of Experimental Abdominal Aortic Aneurysm.
Journal of the American Heart Association (2022-01-06)
Chongyang Zhang, Amy Mohan, Hangchuan Shi, Chen Yan
ABSTRACT
Background cGMP-hydrolyzing phosphodiesterase type 5 (PDE5) regulates vascular smooth muscle cell (SMC) contraction by antagonizing cGMP-dependent protein kinase I (PKGI)-dependent SMC relaxation. SMC contractile dysfunction is implicated in the pathogenesis of aortic aneurysm. PDE5 inhibitors have been used for treating erectile dysfunction, such as drug Viagra (sildenafil). However, a few clinical cases have reported the association of Viagra usage with aortic dissection, and reduced PDE5A expression was found in human aortic aneurysm tissues. Therefore, we aimed to investigate the effect of sildenafil on experimental abdominal aortic aneurysm (AAA), the most common form of aortic aneurysm in elderly men. Methods and Results AAA was induced in C57BL/6J male mice by periaortic elastase in combination with blocking elastin/collagen formation via 3-aminopropionitrile fumarate salt for 35 days. PDE5A protein levels detected by immunostaining were significantly reduced in mouse AAA. Sildenafil application in drinking water significantly aggravated aortic wall dilation and elastin degradation with pre-existing moderate AAA. The phosphorylation level of myosin light chain 2 at Ser19, a biochemical marker of SMC contraction, was significantly reduced by sildenafil in AAA. Proximity ligation assay further revealed that the interaction between cGMP and PKGI was significantly increased by sildenafil in AAA, suggesting an elevation of PKGI activation in AAA. Conclusions Sildenafil treatment aggravated the degradation of elastin fibers and progression of experimental AAA by dysregulating cGMP and contractile signaling in SMCs. Our findings may raise the caution of clinical usage of Viagra in aneurysmal patients.
Sigma-Aldrich
Anti-MMP-9 Antibody, Catalytic domain, Chemicon®, from rabbit
Sigma-Aldrich
Anti-cGMP (cyclic GMP) Antibody, serum, Upstate®
Sigma-Aldrich
Elastase from porcine pancreas, Type I, ≥4.0 units/mg protein