Merck
  • NMR waterLOGSY as An Assay in Drug Development Programmes for Detecting Protein-Ligand Interactions-NMR waterLOGSY.
NMR waterLOGSY as An Assay in Drug Development Programmes for Detecting Protein-Ligand Interactions-NMR waterLOGSY.
Bio-protocol (2021-03-05)
Carole J R Bataille, Terence H Rabbitts, Timothy D W Claridge
ABSTRACT
In drug development programmes, multiple assays are needed for the determination of protein-compound interactions and evaluation of potential use in assays with protein-protein interactions. In this protocol we describe the waterLOGSY NMR method for confirming protein-ligand binding events.
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D