Merck
  • Investigation of Active Anti-Inflammatory Constituents of Essential Oil from Pinus koraiensis (Sieb. et Zucc.) Wood in LPS-Stimulated RBL-2H3 Cells.
Investigation of Active Anti-Inflammatory Constituents of Essential Oil from Pinus koraiensis (Sieb. et Zucc.) Wood in LPS-Stimulated RBL-2H3 Cells.
Biomolecules (2021-06-03)
Jiyoon Yang, Won-Sil Choi, Ki-Joong Kim, Chang-Deuk Eom, Mi-Jin Park
ABSTRACT
In a previous study, we demonstrated the anti-inflammatory activity of the essential oil extracted from Korean pine (Pinus koraiensis, Sieb. et Zucc.) wood. This study aims to investigate the active anti-inflammatory constituents of P. koraiensis oil. The essential oil was extracted from P. koraiensis wood by hydrodistillation and was divided into six fractions (A-F) through fractional distillation. Then, the anti-inflammatory activities of the fractions (A-F) were determined. Fractions A and F markedly downregulated the production of pro-inflammatory cytokines as well as the secretion of β-hexosaminidase in lipopolysaccharide (LPS)-stimulated RBL-2H3 cells. The main constituents of the active anti-inflammatory A and F fractions were (+)-α-pinene, (-)-β-pinene, (+)-α-terpineol, 3-carene, (+)-limonene, and longifolene. These six single compounds decreased the expression of inflammatory-related genes (i.e., IL-4 and IL-13) as well as the secretion of β-hexosaminidase in LPS-stimulated RBL-2H3 cells. (+)-α-Pinene, (-)-β-pinene, (+)-α-terpineol, and longifolene exhibited the strongest effects; these effects were comparable to those of the positive control (i.e., dexamethasone). The findings indicate that the interactions between these components exhibit potential for the management and/or treatment of inflammatory conditions as well as base structures for the development of novel anti-inflammatory drugs.
Sigma-Aldrich
(−)-β-Pinene, ≥99%