Merck
RCAN1 Regulates Bidirectional Synaptic Plasticity.
Current biology : CB (2020-02-23)
Anthony Dudilot, Emilie Trillaud-Doppia, Jannic Boehm
ABSTRACT
Synaptic plasticity, with its two most studied forms, long-term potentiation (LTP) and long-term depression (LTD), is the cellular mechanism underlying learning and memory. Although it has been known for two decades that bidirectional synaptic plasticity necessitates a corresponding bidirectional regulation of calcineurin activity, the underlying molecular mechanism remains elusive. Using organotypic hippocampal slice cultures, we show here that phosphorylation of the endogenous regulator-of-calcineurin (RCAN1) by GSK3β underlies calcineurin activation and is a necessary event for LTD induction, while phosphorylation of RCAN1 at a PKA site blocks calcineurin activity, thereby allowing LTP induction. Our results provide a new mechanism for the regulation of calcineurin in bidirectional synaptic plasticity and establish RCAN1 as a "switch" for bidirectional synaptic plasticity.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Minimum Essential Medium Eagle, With Hanks′ salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Earle′s Balanced Salts, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture