Merck
  • Single-Cell Mapping of Human Brain Cancer Reveals Tumor-Specific Instruction of Tissue-Invading Leukocytes.
Single-Cell Mapping of Human Brain Cancer Reveals Tumor-Specific Instruction of Tissue-Invading Leukocytes.
Cell (2020-05-30)
Ekaterina Friebel, Konstantina Kapolou, Susanne Unger, Nicolás Gonzalo Núñez, Sebastian Utz, Elisabeth Jane Rushing, Luca Regli, Michael Weller, Melanie Greter, Sonia Tugues, Marian Christoph Neidert, Burkhard Becher
ABSTRACT
Brain malignancies can either originate from within the CNS (gliomas) or invade from other locations in the body (metastases). A highly immunosuppressive tumor microenvironment (TME) influences brain tumor outgrowth. Whether the TME is predominantly shaped by the CNS micromilieu or by the malignancy itself is unknown, as is the diversity, origin, and function of CNS tumor-associated macrophages (TAMs). Here, we have mapped the leukocyte landscape of brain tumors using high-dimensional single-cell profiling (CyTOF). The heterogeneous composition of tissue-resident and invading immune cells within the TME alone permitted a clear distinction between gliomas and brain metastases (BrM). The glioma TME presented predominantly with tissue-resident, reactive microglia, whereas tissue-invading leukocytes accumulated in BrM. Tissue-invading TAMs showed a distinctive signature trajectory, revealing tumor-driven instruction along with contrasting lymphocyte activation and exhaustion. Defining the specific immunological signature of brain tumors can facilitate the rational design of targeted immunotherapy strategies.
Product Description
Hamilton® 75N syringe, 75N, volume 5 μL, needle size 26s ga (cone tip), needle L 51 mm (2 in.)
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
Saponin from quillaja bark, Sapogenin content ≥10 %