Merck
  • Enhanced docetaxel delivery using sterically stabilized RIPL peptide-conjugated nanostructured lipid carriers: In vitro and in vivo antitumor efficacy against SKOV3 ovarian cancer cells.
Enhanced docetaxel delivery using sterically stabilized RIPL peptide-conjugated nanostructured lipid carriers: In vitro and in vivo antitumor efficacy against SKOV3 ovarian cancer cells.
International journal of pharmaceutics (2020-05-08)
Chang Hyun Kim, Tae Hoon Kang, Byoung Deok Kim, Tae Hwa Lee, Ho Yub Yoon, Yoon Tae Goo, Yong Seok Choi, Myung Joo Kang, Young Wook Choi
ABSTRACT
Docetaxel (DTX) has poor solubility, low specificity, and severe side effects. For efficient targeting of DTX to hepsin-overexpressing SKOV3 ovarian cancer cells, PEGylated and RIPL peptide (IPLVVPLRRRRRRRRC)-conjugated nanostructured lipid carriers (PEG-RIPL-NLCs) were examined for in vitro and in vivo antitumor efficacy. DTX-loaded plain NLCs (DTX-pNLCs), RIPL-NLCs (DTX-RIPL-NLCs), and PEG-RIPL-NLCs (DTX-PEG-RIPL-NLCs) were prepared using a solvent emulsification-evaporation technique. DTX was successfully loaded with high encapsulation efficiency (>93%), and all NLCs showed homogeneous dispersion with zeta potentials varying from -17 to 15 mV. Drug release was biphasic: initial rapid release, then gradual release. In vitro cytotoxicity was time- and dose-dependent: DTX-RIPL-NLCs and DTX-PEG-RIPL-NLCs exhibited greater cytotoxicity, enhanced cell apoptosis owing to the cell cycle arrest in the G2/M phase, and increased activation of the mitochondria-related intrinsic apoptosis pathway compared to DTX-pNLCs. Pharmacokinetic experiments in male Sprague-Dawley rats revealed that DTX-PEG-RIPL-NLCs increased the mean residence time of DTX but reduced total body clearance and volume of distribution. In a SKOV3-bearing xenograft Balb/c athymic mouse model, DTX-PEG-RIPL-NLCs suppressed tumors, evidenced by tumor volume change and histopathological examination. Thus, we conclude that PEG-RIPL-NLCs have an advantage of high payload of poorly water-soluble drugs and are a good candidate for drug targeting to SKOV3-derived ovarian cancer.
Product Description
Avanti
DSPE-PEG(2000) Maleimide, Avanti Research - A Croda Brand 880126C
Avanti
DSPE-PEG(2000) Maleimide, Avanti Research - A Croda Brand
Avanti
18:0 PEG3000 PE, Avanti Research - A Croda Brand 880320P, powder
Avanti
18:0 PEG3000 PE, Avanti Research - A Croda Brand 880320C