- Flavor Preference and Systemic Immunoglobulin Responses in E-Cigarette Users and Waterpipe and Tobacco Smokers: A Pilot Study.
Flavor Preference and Systemic Immunoglobulin Responses in E-Cigarette Users and Waterpipe and Tobacco Smokers: A Pilot Study.
Electronic cigarette (e-cigarette) use has had an exponential increase in popularity since the product was released to the public. Currently, there is a lack of human studies that assess different biomarker levels. This pilot study attempts to link e-cigarette and other tobacco product usage with clinical respiratory symptoms and immunoglobulin response. Subjects completed surveys in order to collect self-reported data on tobacco product flavor preferences. Along with this, plasma samples were collected to test for immunoglobulin G (IgG) and E (IgE) levels. Our pilot study's cohort had a 47.9% flavor preference towards fruit flavors and a 63.1% preference to more sweet flavors. E-cigarette and traditional cigarette smokers were the two subject groups to report the most clinical symptoms. E-cigarette users also had a significant increase in plasma IgE levels compared to non-tobacco users 1, and dual users had a significant increase in plasma IgG compared to non-tobacco users 2, cigarette smokers, and waterpipe smokers. Our pilot study showed that users have a preference toward fruit and more sweet flavors and that e-cigarette and dual use resulted in an augmented systemic immune response.