Merck
  • Light-Activated Rhenium Complexes with Dual Mode of Action against Bacteria.
Light-Activated Rhenium Complexes with Dual Mode of Action against Bacteria.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-12-04)
Angelo Frei, Maite Amado, Matthew A Cooper, Mark A T Blaskovich
ABSTRACT
New antibiotics and innovative approaches to kill drug-resistant bacteria are urgently needed. Metal complexes offer access to alternative modes of action but have only sparingly been investigated in antibacterial drug discovery. We have developed a light-activated rhenium complex with activity against drug-resistant S. aureus and E. coli. The activity profile against mutant strains combined with assessments of cellular uptake and synergy suggest two distinct modes of action.
Sigma-Aldrich
Gentamicin sulfate, meets USP testing specifications, powder
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications
Meropenem trihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Millipore
MRSA Selective Supplement, suitable for microbiology