Merck
  • Prolactin stimulates prostate cell proliferation by increasing endoplasmic reticulum content due to SERCA 2b over-expression.
Prolactin stimulates prostate cell proliferation by increasing endoplasmic reticulum content due to SERCA 2b over-expression.
The Biochemical journal (2006-09-13)
Alexandre Crépin, Gabriel Bidaux, Fabien Vanden-Abeele, Etienne Dewailly, Vincent Goffin, Natalia Prevarskaya, Christian Slomianny
ABSTRACT
Prolactin (PRL) has been shown to be involved in the differentiation and proliferation of numerous tissues, including the prostate gland. Moreover, variations in [Ca2+]ER (calcium concentration within the endoplasmic reticulum) may play a role in cell growth. However, few studies have focused on the regulation of calcium homoeostasis by prolactin. The present study evaluates the regulation of calcium pools as well as the possible role of [Ca2+]ER variations as a signal for growth modulation by PRL. We show that PRL stimulates the proliferation of normal SV40 immortalized epithelial prostate (PNT1A) cells with a maximum effect at a dose of 100 ng/ml. We also show that 100 ng/ml PRL increases the [Ca2+]ER when measured either by indirect quantification with Fura-2AM after application of 1 mM thapsigargin or by direct quantification with Mag-Fura-2AM within the endoplas-mic reticulum. Western blot analysis shows that the SERCA 2b (sarcoendoplasmic calcium ATPase 2b) is over-expressed in PNT1A cells treated with 100 ng/ml PRL for 24 h. A small inter-fering RNA SERCA 2a/b, used to down-regulate endogenous SERCA 2b expression, reduced both PNT1A cell proliferation and [Ca2+]ER. We thus identify [Ca2+]ER and SERCA 2b as protagonists in PRL-induced proliferation.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey