
Zopiclone oxide

Synonym(s):
(5RS)-6-(5-Chloropyridin-2-yl)-7-oxo-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-yl 4-methylpiperazine-1-carboxylate 4-oxide
Empirical Formula (Hill Notation):
C17H17ClN6O4
CAS Number:
Molecular Weight:
404.81
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing