
Vinorelbine for peak identification B

Synonym(s):
Vinorelbine ditartrate salt
Empirical Formula (Hill Notation):
C45H54N4O8 · 2C4H6O6
CAS Number:
Molecular Weight:
1079.11
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing