
Trigonelline chloride

Synonym(s):
Trigonelline hydrochloride, 1-Methylpyridinium-3-carboxylate hydrochloride, N-Methylnicotinic acid betaine hydrochloride
Empirical Formula (Hill Notation):
C7H7NO2 · HCl
CAS Number:
Molecular Weight:
173.60
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing