Skip to Content
Merck
Tributylmethylammonium chloride ≥98.0% (T)

Tributylmethylammonium chloride

Tributylmethylammonium chloride ≥98.0% (T)
Synonym(s):
Methyltributylammonium chloride
Linear Formula:
(CH3CH2CH2CH2)3N(Cl)CH3
CAS Number:
Molecular Weight:
235.84
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing