
Sotalol Related Compound A

Synonym(s):
N-{4-{[(1-Methylethyl)amino]acetyl}phenyl}methanesulfonamid hydrochloride
Empirical Formula (Hill Notation):
C12H18N2O3S · HCl
CAS Number:
Molecular Weight:
306.81
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2