
(S)-Naproxen methyl ester

Synonym(s):
(2S)-2-(6-Methoxy-2-naphthyl)propionic acid methyl ester, Methyl (αS)-6-methoxy-α-methyl-2-naphthaleneacetate, Methyl (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
Empirical Formula (Hill Notation):
C15H16O3
CAS Number:
Molecular Weight:
244.29
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing