
RRR-α-Tocopheryl hydrogen succinate

Synonym(s):
D-α-Tocopherol succinate, Vitamin E succinate
Empirical Formula (Hill Notation):
C33H54O5
CAS Number:
Molecular Weight:
530.78
Products
Product Category
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing