
Pyrantel impurity C

Synonym(s):
2-Thiophenecarboxaldehyde, Thenaldehyde
Empirical Formula (Hill Notation):
C5H4OS
CAS Number:
Molecular Weight:
112.15
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2