
Proteinase K from Tritirachium album

Synonym(s):
Endopeptidase K, Proteinase K from Tritirachium album
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing