
Pinoresinol diglucoside

Synonym(s):
(+)-Pinoresinol 4,4′-O-bisglucopyranoside, Pinoresinol di-β-D-glucoside
Empirical Formula (Hill Notation):
C32H42O16
CAS Number:
Molecular Weight:
682.67
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing