
O-(Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate

Synonym(s):
N,N,N′,N′-Tetramethyl-O-(benzotriazol-1-yl)uronium tetrafluoroborate, TBTU
Linear Formula:
C11H16N5O · BF4
CAS Number:
Molecular Weight:
321.08
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing