
N-(2-Hydroxypropyl)-2-methyl-prop-2-enamide

Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing