
Monoclonal Anti-Cytokeratin, pan antibody produced in mouse

Synonym(s):
Monoclonal Anti-Pan Cytokeratin, Monoclonal Anti-pan-Cytokeratin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2